4-Methylbenzamidin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 4-Methylbenzoesäureamidin Hydrochlorid, 4-Methylphenylamidin Hydrochlorid

12-0030.1 12-0030.5
CHMP12-0030.1EA 0 EUR
CHMP12-0030.1 CHMP12-0030.5
4-Methylbenzamidin Hydrochlorid
4-Methylbenzamidin Hydrochlorid
Formel: C₈H₁₁ClN₂
Molecular Weight: 170,64 g/mol
MDL: MFCD00040183
CAS-Nummer: 6326-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR