4-Tolylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Methylphenyl)metaborsäure, 4-Methylphenylboronsäure, 4-Methylbenzolboronsäure, Dihydroxy(4-methylphenyl)boran
BB-2337-5G
BB-2337-100G
BB-2337-25G
COBBBB-2337-5GEA
25.5
EUR
COBBBB-2337-5G
COBBBB-2337-100G
COBBBB-2337-25G
4-Tolylboronsäure
4-Tolylboronsäure
Formel:
C₇H₉BO₂ Molecular Weight: 135,96 g/mol Schmelzpunkt: 260 °C |
MDL:
MFCD00039138 CAS-Nummer: 5720-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...