4-Nitrobenzolsulfonamid ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Nitrobenzolsulfonsäureamid

A18101.06 A18101.14
A18101.06EA 48.1 EUR
A18101.06 A18101.14
4-Nitrobenzolsulfonamid ≥97%
4-Nitrobenzolsulfonamid

Purity: 97% CAS No.: 6325-93-5 Molecular Formula: C6H6N2O4S Formula Weight: 202.19 Melting Point: 178-181° MDL No.: MFCD00007937

Formel: C₆H₆N₂O₄S
Molecular Weight: 202,19 g/mol
Schmelzpunkt: 178…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007937
CAS-Nummer: 6325-93-5
EINECS: 228-691-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR