4-Nitrobenzolsulfonamid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Nitrobenzolsulfonsäureamid

153020050 153020250
ACRO153020050EA 21.1 EUR
ACRO153020050 ACRO153020250
4-Nitrobenzolsulfonamid 97%
4-Nitrobenzolsulfonamid
Formel: C₆H₆N₂O₄S
Molecular Weight: 202,19 g/mol
Schmelzpunkt: 178…182 °C
MDL: MFCD00007937
CAS-Nummer: 6325-93-5
EINECS: 228-691-6

Bestellen

Spezifikation Testergebnisse

Appearance White to beige to tan Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 175 to 181 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR