4-Nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Nitrophenyl)metaborsäure, 4-Nitrobenzolboronsäure, Dihydroxy(4-nitrophenyl)boran

BB-2450-5G BB-2450-1G BB-2450-25G
COBBBB-2450-5GEA 169 EUR
COBBBB-2450-5G COBBBB-2450-1G COBBBB-2450-25G
4-Nitrophenylboronsäure
4-Nitrophenylboronsäure
Formel: C₆H₆BNO₄
Molecular Weight: 166.93 g/mol
MDL: MFCD00161360
CAS-Nummer: 24067-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR