4-Pentylbenzolsulfonylchlorid
Lieferant: MENAI
Synonyme:
4-Amylbenzol-1-sulfonylchlorid, 4-Pentylbenzolsulfonsäurechlorid, 4-Pentylbenzolsulfochlorid, 4-Amylbenzolsulfochlorid, 4-Pentylbenzol-1-sulfonylchlorid, 4-Amylbenzolsulfonsäurechlorid, 4-Amylbenzolsulfonylchlorid
E3679.50
E3679.25
MENAE3679.50EA
0
EUR
MENAE3679.50
MENAE3679.25
4-Pentylbenzolsulfonylchlorid
4-Pentylbenzolsulfonylchlorid
Formel:
C₁₁H₁₅ClO₂S Molecular Weight: 246.76 g/mol |
MDL:
MFCD00173759 CAS-Nummer: 73948-18-2 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...