4-Pentylbenzolsulfonylchlorid ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Amylbenzol-1-sulfonylchlorid, 4-Pentylbenzolsulfonsäurechlorid, 4-Pentylbenzolsulfochlorid, 4-Amylbenzolsulfochlorid, 4-Pentylbenzol-1-sulfonylchlorid, 4-Amylbenzolsulfonsäurechlorid, 4-Amylbenzolsulfonylchlorid
A19929.88
A19929.09
A19929.88EA
29.3
EUR
A19929.88
A19929.09
4-Pentylbenzolsulfonylchlorid ≥97%
4-Pentylbenzolsulfonylchlorid
Purity: 97%
CAS No.: 73948-18-2
Molecular Formula: C11H15ClO2S
Formula Weight: 246.76
Melting Point: 39-44°
UN No.: 3261
MDL No.: MFCD00173759
Formel:
C₁₁H₁₅ClO₂S Molecular Weight: 246,76 g/mol Schmelzpunkt: 39…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173759 CAS-Nummer: 73948-18-2 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...