5-Brom-1-indanon

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2,3-dihydro-1H-inden-1-one

OS-7501-25G OS-7501-100G OS-7501-5G
COBBOS-7501-25GEA 95.9 EUR
COBBOS-7501-25G COBBOS-7501-100G COBBOS-7501-5G
5-Brom-1-indanon
5-Brom-1-indanon
Formel: C₉H₇BrO
Molecular Weight: 211,06 g/mol
Siedepunkt: 280 °C (1013 hPa)
Schmelzpunkt: 126…127 °C
MDL: MFCD00082718
CAS-Nummer: 34598-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR