5-Brom-1-methylimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
5-Brom-1-methyl-1H-imidazol
HI-1025-1G
HI-1025-5G
HI-1025-100G
HI-1025-25G
COBBHI-1025-1GEA
0
EUR
COBBHI-1025-1G
COBBHI-1025-5G
COBBHI-1025-100G
COBBHI-1025-25G
5-Brom-1-methylimidazol
5-Bromo-1-methylimidazole
Formel:
C₄H₅BrN₂ Molecular Weight: 161 g/mol |
MDL:
MFCD01632218 CAS-Nummer: 1003-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...