5-Brom-1-methylimidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Brom-1-methyl-1H-imidazol
M-Clarity™ Qualitätsstufe = MQ100
568279-25G
568279-5G
SIAL568279-25GEA
806
EUR
SIAL568279-25G
SIAL568279-5G
5-Brom-1-methylimidazol, Sigma-Aldrich®
5-Bromo-1-methylimidazole
Formel:
C₄H₅BrN₂ Molecular Weight: 161 g/mol |
MDL:
MFCD01632218 CAS-Nummer: 1003-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...