5-Brom-2,4-dichlorpyrimidin
Lieferant: COMBI-BLOCKS
OS-7567-5G
OS-7567-25G
OS-7567-100G
COBBOS-7567-5GEA
0
EUR
COBBOS-7567-5G
COBBOS-7567-25G
COBBOS-7567-100G
5-Brom-2,4-dichlorpyrimidin
5-Bromo-2,4-dichloropyrimidine
Formel:
C₄HBrCl₂N₂ Molecular Weight: 227.87 g/mol Siedepunkt: 288 Schmelzpunkt: –1 °C Dichte: 1.781 g/cm³ (20 °C) |
MDL:
MFCD00127818 CAS-Nummer: 36082-50-5 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...