5-Brom-2-chlornicotinsäure

Lieferant: COMBI-BLOCKS

CA-4175-5G CA-4175-25G
COBBCA-4175-5GEA 26.2 EUR
COBBCA-4175-5G COBBCA-4175-25G
5-Brom-2-chlornicotinsäure
5-Brom-2-chlornicotinsäure
Formel: C₆H₃BrClNO₂
MDL: MFCD03844847
CAS-Nummer: 29241-65-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR