5-Brom-2-chlorpyrimidin

Lieferant: COMBI-BLOCKS

OS-7523-25G OS-7523-5G OS-7523-100G
COBBOS-7523-25GEA 63.4 EUR
COBBOS-7523-25G COBBOS-7523-5G COBBOS-7523-100G
5-Brom-2-chlorpyrimidin
5-Bromo-2-chloropyrimidine
Formel: C₄H₂BrClN₂
Molecular Weight: 193,43 g/mol
MDL: MFCD00483232
CAS-Nummer: 32779-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR