5-Brom-2-thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromthiophen-2-boronsäure, 5-Brom-2-thienylboronsäure

BB-3866-5G BB-3866-25G BB-3866-1G
COBBBB-3866-5GEA 102 EUR
COBBBB-3866-5G COBBBB-3866-25G COBBBB-3866-1G
5-Brom-2-thiophenboronsäure
5-Bromo-2-thiopheneboronic acid
Formel: C₄H₄BBrO₂S
Molecular Weight: 206,85 g/mol
MDL: MFCD02094028
CAS-Nummer: 162607-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR