5-Chlor-2-thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Chlor-2-thienylboronsäure, 5-Chlorthiophen-2-boronsäure

BB-2376-5G BB-2376-25G BB-2376-1G
COBBBB-2376-5GEA 142 EUR
COBBBB-2376-5G COBBBB-2376-25G COBBBB-2376-1G
5-Chlor-2-thiophenboronsäure
5-Chloro-2-thiopheneboronic acid
Formel: C₄H₄BClO₂S
Molecular Weight: 162,4 g/mol
MDL: MFCD00236030
CAS-Nummer: 162607-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR