5-Hydroxynicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Hydroxypyridin-3-carbonsäure, 5-Hydroxy-3-pyridincarbonsäure

CA-4059-100G CA-4059-5G CA-4059-25G
COBBCA-4059-100GEA 312 EUR
COBBCA-4059-100G COBBCA-4059-5G COBBCA-4059-25G
5-Hydroxynicotinsäure
5-Hydroxynicotinsäure
Formel: C₆H₅NO₃
Molecular Weight: 139,11 g/mol
MDL: MFCD00129117
CAS-Nummer: 27828-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR