5-Indolcarbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
1H-Indole-5-carboxylic acid
IN-0011-5G
IN-0011-25G
IN-0011-1G
COBBIN-0011-5GEA
0
EUR
COBBIN-0011-5G
COBBIN-0011-25G
COBBIN-0011-1G
5-Indolcarbonsäure
5-Indolcarbonsäure
Formel:
C₉H₇NO₂ Schmelzpunkt: 211…213 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005678 CAS-Nummer: 1670-81-1 EINECS: 216-799-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...