5-Methoxyindol
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
5-Methoxy-1H-indole, Indol-5-yl methyl ether
IN-0049-5G
IN-0049-25G
IN-0049-100G
COBBIN-0049-5GEA
61.3
EUR
COBBIN-0049-5G
COBBIN-0049-25G
COBBIN-0049-100G
5-Methoxyindol
5-Methoxyindol
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol Siedepunkt: 312 °C (1013 hPa) Schmelzpunkt: 55…56 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005674 CAS-Nummer: 1006-94-6 EINECS: 213-745-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...