5-Methylnicotinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
5-Methyl-3-pyridincarbonsäure, 5-Methylpyridin-3-carbonsäure
PY-1457-100G
PY-1457-25G
PY-1457-5G
COBBPY-1457-100GEA
175
EUR
COBBPY-1457-100G
COBBPY-1457-25G
COBBPY-1457-5G
5-Methylnicotinsäure
5-Methylnicotinsäure
Formel:
C₇H₇NO₂ Molecular Weight: 137.14 g/mol |
MDL:
MFCD00829036 CAS-Nummer: 3222-49-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...