5-Methylchinoxalin

Lieferant: COMBI-BLOCKS
Achtung

HC-3121-5G HC-3121-25G HC-3121-1G
COBBHC-3121-5GEA 51.3 EUR
COBBHC-3121-5G COBBHC-3121-25G COBBHC-3121-1G
5-Methylchinoxalin
5-Methylquinoxaline
Formel: C₉H₈N₂
Molecular Weight: 144.18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00012335
CAS-Nummer: 13708-12-8
EINECS: 237-246-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR