2-Brom-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-5-pyridinylboronsäure, 2-Brompyridin-5-boronsäure

BB-3423-5G BB-3423-25G BB-3423-1G
COBBBB-3423-5GEA 73.1 EUR
COBBBB-3423-5G COBBBB-3423-25G COBBBB-3423-1G
2-Brom-5-pyridinboronsäure
2-Brom-5-pyridinboronsäure
Formel: C₅H₅BBrNO₂
Molecular Weight: 201,81 g/mol
MDL: MFCD03411558
CAS-Nummer: 223463-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR