2-Fluor-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluorpyridin-5-boronsäure, 2-Fluor-5-pyridinylboronsäure

BB-3448-5G BB-3448-25G BB-3448-1G BB-3448-100G
COBBBB-3448-5GEA 51.3 EUR
COBBBB-3448-5G COBBBB-3448-25G COBBBB-3448-1G COBBBB-3448-100G
2-Fluor-5-pyridinboronsäure
2-Fluor-5-pyridinboronsäure
Formel: C₅H₅BFNO₂
Molecular Weight: 140,91 g/mol
MDL: MFCD03411559
CAS-Nummer: 351019-18-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR