7-Fluorisatin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
7-Fluorindolin-2,3-dion, 7-Fluor-2,3-indolindion, 7-Fluor-2,3-dioxoindolin, 7-Fluor-1H-indol-2,3-dion
IN-0130-100G
IN-0130-5G
IN-0130-25G
COBBIN-0130-100GEA
451
EUR
COBBIN-0130-100G
COBBIN-0130-5G
COBBIN-0130-25G
7-Fluorisatin
7-Fluorisatin
Formel:
C₈H₄FNO₂ Molecular Weight: 165,12 g/mol |
MDL:
MFCD01569508 CAS-Nummer: 317-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...