6-Indolcarbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1H-Indole-6-carboxylic acid

IN-0018-100G IN-0018-25G IN-0018-5G
COBBIN-0018-100GEA 0 EUR
COBBIN-0018-100G COBBIN-0018-25G COBBIN-0018-5G
6-Indolcarbonsäure
6-Indolcarbonsäure
Formel: C₉H₇NO₂
Molecular Weight: 161.16 g/mol
Schmelzpunkt: 254…257 °C
MDL: MFCD00210441
CAS-Nummer: 1670-82-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR