6-Methylchinoxalin
Lieferant: COMBI-BLOCKS
|
Achtung
|
HC-3164-5G
HC-3164-25G
HC-3164-1G
COBBHC-3164-5GEA
67.3
EUR
COBBHC-3164-5G
COBBHC-3164-25G
COBBHC-3164-1G
6-Methylchinoxalin
6-Methylquinoxaline
Formel:
C₉H₈N₂ Molecular Weight: 144.18 g/mol |
MDL:
MFCD00041001 CAS-Nummer: 6344-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...