7-Aminoindol
Lieferant: COMBI-BLOCKS
Synonyme:
Indol-7-ylamin, 7-Amino-1H-indol, 7-Indolamin
IN-0027-5G
IN-0027-1G
COBBIN-0027-5GEA
802
EUR
COBBIN-0027-5G
COBBIN-0027-1G
7-Aminoindol
7-Aminoindol
Formel:
C₈H₈N₂ Molecular Weight: 132,16 g/mol Schmelzpunkt: 99…199 °C |
MDL:
MFCD02093960 CAS-Nummer: 5192-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...