7-Chlorindol

Lieferant: COMBI-BLOCKS
Achtung

IN-0059-5G IN-0059-25G IN-0059-1G IN-0059-100G
COBBIN-0059-5GEA 159 EUR
COBBIN-0059-5G COBBIN-0059-25G COBBIN-0059-1G COBBIN-0059-100G
7-Chlorindol
7-Chlorindol
Formel: C₈H₆ClN
Molecular Weight: 151.6 g/mol
MDL: MFCD01321301
CAS-Nummer: 53924-05-3
EINECS: 258-865-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR