8-Bromchinolin

Lieferant: COMBI-BLOCKS
Achtung

PY-7342-25G PY-7342-5G
COBBPY-7342-25GEA 138 EUR
COBBPY-7342-25G COBBPY-7342-5G
8-Bromchinolin
8-Bromoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD00191859
CAS-Nummer: 16567-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR