Diammoniumhexabromoosmat (≥26,5% Os) ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ammoniumhexabromoosmat(IV)
12648.03
12648.06
12648.03EA
129
EUR
12648.03
12648.06
Diammoniumhexabromoosmat (≥26,5% Os) ≥99,9% (Metall-Basis)
Diammoniumhexabromoosmat
Purity: 99.9%
CAS No.: 24598-62-7
Molecular Formula: (NH4)2OsBr6
Formula Weight: 705.73
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Black
Odour: Odourless
MDL No.: MFCD00049389
Formel:
H₈Br₆N₂Os Molecular Weight: 705,73 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049389 CAS-Nummer: 24598-62-7 EINECS: 246-340-5 |