Ammoniumhexachloroosmat(IV) ≥99,99% (Metall-Basis) Os ≥ 42.8%, Premion®
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Diammoniumhexachloroosmat
10823.02
10823.03
10823.06
10823.02EA
121
EUR
10823.02
10823.03
10823.06
Ammoniumhexachloroosmat(IV) ≥99,99% (Metall-Basis) Os ≥ 42.8%, Premion®
Ammonium hexachloroosmate(IV)
Purity: 99.99%
CAS No.: 12125-08-5
Molecular Formula: H8Cl6N2Os
Formula Weight: 438.99
Physical Form: Powder
Density: 2.9
Appearance: Brown to black
Melting Point: 170° subl.
Merck Reference: 14,536
MDL No.: MFCD00010883
Formel:
H₈Cl₆N₂Os Molecular Weight: 439,02 g/mol Schmelzpunkt: 170 °C (Subl.) Dichte: 2,9 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010883 CAS-Nummer: 12125-08-5 EINECS: 235-188-5 Merck Index: 14,536 |