Benzamidin Hydrochlorid Hydrat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Phenylamidin Hydrochlorid Hydrat, Benzoesäureamidin Hydrochlorid Hydrat, Benzamidiniumchlorid Hydrat
M-Clarity™ Qualitätsstufe = MQ200
B6506-100G
B6506-25G
B6506-5G
SIALB6506-100GEA
354
EUR
SIALB6506-100G
SIALB6506-25G
SIALB6506-5G
Benzamidin Hydrochlorid Hydrat, Sigma-Aldrich®
Benzamidin Hydrochlorid Hydrat
Formel:
C₆H₅C(=NH)NH₂·HCl·H₂O Molecular Weight: 174,63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066285 CAS-Nummer: 206752-36-5 EINECS: 216-795-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...