Bilirubin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(2-((3-(2-Carboxyethyl)-4-methyl-5-((Z)-(3-methyl-5-oxo-4-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-2-yl)methyl)-4-methyl-5-((Z)-(4-methyl-5-oxo-3-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid

14370-250MG 14370-1G 14370-5G
SIAL14370-250MGEA 87 EUR
SIAL14370-250MG SIAL14370-1G SIAL14370-5G
Bilirubin, Sigma-Aldrich®
Bilirubin
Formel: C₃₃H₃₆N₄O₆
Molecular Weight: 584.67 g/mol
Schmelzpunkt: > 360 °C
Dichte: 1.31 g/cm³ (20 °C)
MDL: MFCD00005499
CAS-Nummer: 635-65-4
EINECS: 211-239-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR