Bromcyclobutan
Lieferant: COMBI-BLOCKS
|
Gefahr
|
Synonyme:
Cyclobutylbromid
OS-7568-5G
OS-7568-25G
OS-7568-1G
COBBOS-7568-5GEA
98.3
EUR
COBBOS-7568-5G
COBBOS-7568-25G
COBBOS-7568-1G
Bromcyclobutan
Bromcyclobutan
Formel:
C₄H₇Br Molecular Weight: 135 g/mol Siedepunkt: 109…113 °C (1013 hPa) Dichte: 1,4208 g/cm³ (20 °C) |
MDL:
MFCD00001317 CAS-Nummer: 4399-47-7 EINECS: 224-530-9 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...