Carbazol 95%

Lieferant: Thermo Scientific
Achtung

A11448.18 A11448.22 A11448.36
A11448.18EA 41.4 EUR
A11448.18 A11448.22 A11448.36
Carbazol 95%
Carbazole

Purity: 95% CAS No.: 86-74-8 Molecular Formula: C12H9N Formula Weight: 167.21 Flash Point: 220°(428°F) Odour: Odourless Boiling Point: 354-356° Beilstein Reference: 3956 Merck Reference: 14,1790 UN No.: 3077 MDL No.: MFCD00004960

Formel: C₁₂H₉N
Molecular Weight: 167,21 g/mol
Siedepunkt: 354…356 °C
Schmelzpunkt: 240…246 °C
Dichte: 1,150
Flammpunkt: 220 °C (428 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004960
CAS-Nummer: 86-74-8
EINECS: 201-696-0
ADR: 9,III
Merck Index: 14,1790

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR