Benzylcyanacetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cyanessigsäurebenzylester, Phenylmethylcyanacetat

663824-25G 663824-5G
SIAL663824-25GEA 118 EUR
SIAL663824-25G SIAL663824-5G
Benzylcyanacetat, Sigma-Aldrich®
Benzylcyanacetat
Formel: C₁₀H₉NO₂
Molecular Weight: 175.19 g/mol
MDL: MFCD00051719
CAS-Nummer: 14447-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR