Cyclobutanon

Lieferant: COMBI-BLOCKS

OS-7558-5G OS-7558-25G
COBBOS-7558-5GEA 38.2 EUR
COBBOS-7558-5G COBBOS-7558-25G
Cyclobutanon
Cyclobutanone
Formel: C₄H₆O
Molecular Weight: 70.09 g/mol
Siedepunkt: 99
Dichte: 0.93 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00001332
CAS-Nummer: 1191-95-3
EINECS: 214-745-6
UN: 1224
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR