Cyclopropyltriphenylphosphoniumbromid

Lieferant: CHEMPUR

CD00857.250 CD00857.1
CHMPCD00857.250EA 0 EUR
CHMPCD00857.250 MAYBCD00857.1
Cyclopropyltriphenylphosphoniumbromid
Cyclopropyltriphenylphosphonium bromide
Formel: C₂₁H₂₀BrP
Molecular Weight: 383,27 g/mol
Schmelzpunkt: 189…190 °C
MDL: MFCD00011872
CAS-Nummer: 14114-05-7
EINECS: 237-970-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR