Cytochalasin B
Lieferant: Serva
Synonyme:
7(S),20(R)-Dihydroxy-16(R)-methyl-10-phenyl-24-oxa[14]cytochalasa-6(12),13(E),21(E)-triene-1,23-dione
18015.01
18015.02
SERA18015.01EA
77.6
EUR
SERA18015.01
SERA18015.02
Cytochalasin B
Cytochalasin B
Formel:
C₂₉H₃₇NO₅ Molecular Weight: 479,62 g/mol Schmelzpunkt: 218…221 °C |
MDL:
MFCD00077704 CAS-Nummer: 14930-96-2 EINECS: 239-000-2 UN: 1544 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...