Cytochalasin D (aus Zygosporium mansoni), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-trien-1,17-dion-21(R)-acetat

M-Clarity™ Qualitätsstufe = MQ300

C8273-10MG C8273-1MG C8273-5MG
SIALC8273-10MGEA 1050 EUR
SIALC8273-10MG SIALC8273-1MG SIALC8273-5MG
Cytochalasin D (aus Zygosporium mansoni), Sigma-Aldrich®
Cytochalasin D
Formel: C₃₀H₃₇NO₆
Molecular Weight: 507,63 g/mol
MDL: MFCD00077706
CAS-Nummer: 22144-77-0
EINECS: 244-804-1
UN: 1544
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR