Dihydrocytochalasin B, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

D1641-1MG D1641-5MG
SIALD1641-1MGEA 350 EUR
SIALD1641-1MG SIALD1641-5MG
Dihydrocytochalasin B, Sigma-Aldrich®
Dihydrocytochalasin B
Formel: C₂₉H₃₉NO₅
Molecular Weight: 481,63 g/mol
Schmelzpunkt: 198…203 °C
CAS-Nummer: 39156-67-7
EINECS: 254-324-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR