Palladium(II)acetat Trimer (≥47% Pd) ≥99,98% (Metall-Basis)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Palladiumdiacetat, Essigsäure Palladium(II)salz
43086.03 43086.06
43086.03EA 173 EUR
43086.03 43086.06
Palladium(II)acetat Trimer (≥47% Pd) ≥99,98% (Metall-Basis)
Palladium(II)acetat
Purity: 99.98% CAS No.: 3375-31-3 Molecular Formula: C12H18O12Pd3 Formula Weight: 673.46 Physical Form: Powder Appearance: Red-brown Melting Point: ca 205° dec. Merck Reference: 14,6991 MDL No.: MFCD00012453
Formel: C₁₂H₁₈O₁₂Pd₃
Molecular Weight: 224,51 g/mol
Schmelzpunkt: ∼ 205 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012453
CAS-Nummer: 3375-31-3
EINECS: 222-164-4
Merck Index: 14,6991