Palladium(II)acetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Palladiumdiacetat
, Essigsäure Palladium(II)salz
M-Clarity™ Qualitätsstufe = MQ200
SIAL379875-1GEA
212
EUR
SIAL379875-1G
SIAL379875-5G
Palladium(II)acetat, Sigma-Aldrich®
Palladium(II)acetat
Formel:
C₄H₆O₄Pd Molecular Weight: 224,51 g/mol Schmelzpunkt: 205 °C |
MDL:
MFCD00012453 CAS-Nummer: 3375-31-3 EINECS: 222-164-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...