2,2,2-Trifluorethylacetat

Lieferant: FLUOROCHEM

Synonyme: Essigsäure(2,2,2-trifluorethyl)ester

007516-25G 007516-100G
FLUO007516-25GEA 82.1 EUR
FLUO007516-25G FLUO007516-100G
2,2,2-Trifluorethylacetat
2,2,2-Trifluorethylacetat
Formel: C₄H₅F₃O₂
Molecular Weight: 142.08 g/mol
Siedepunkt: 77,8
Dichte: 1.2887 g/cm³ (0 °C)
MDL: MFCD00198018
CAS-Nummer: 406-95-1
UN: 3272
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR