2'-Fluoracetanilid

Lieferant: COMBI-BLOCKS

Synonyme: N-(2-Fluorphenyl)acetamid

OT-0027-25G OT-0027-100G
COBBOT-0027-25GEA 45.2 EUR
COBBOT-0027-25G COBBOT-0027-100G
2'-Fluoracetanilid
2'-Fluoracetanilid
Formel: C₈H₈FNO
Molecular Weight: 153,16 g/mol
Siedepunkt: 140…142 °C (14 mmHg)
Schmelzpunkt: 81 °C
MDL: MFCD00000315
CAS-Nummer: 399-31-5
EINECS: 206-916-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR