4-Brombutylacetat

Lieferant: CHEMPUR

Synonyme: Essigsäure(4-brombutyl)ester

RH02076.250 RH02076.1
CHMPRH02076.250EA 0 EUR
CHMPRH02076.250 MAYBRH02076.1
4-Brombutylacetat
4-Brombutylacetat
Formel: C₆H₁₁BrO₂
Molecular Weight: 195,06 g/mol
MDL: MFCD00000263
CAS-Nummer: 4753-59-7
EINECS: 225-277-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR