4-Chlorbutylacetat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Essigsäure(4-chlorbutyl)ester

B22582.14 B22582.22 B22582.36
B22582.14EA 39.9 EUR
B22582.14 B22582.22 B22582.36
4-Chlorbutylacetat ≥98%
4-Chlorbutylacetat

Purity: 98% CAS No.: 6962-92-1 Molecular Formula: CH3CO2(CH2)4Cl Formula Weight: 150.61 Flash Point: 64°(147°F) Refractive Index: 1.4350 Boiling Point: 81-83°/11mm Beilstein Reference: 1749681 MDL No.: MFCD00001013

Formel: C₆H₁₁ClO₂
Molecular Weight: 150.6 g/mol
Siedepunkt: 81…83 °C (11 mmHg)
Dichte: 1.076
Flammpunkt: 64 °C (147 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001013
CAS-Nummer: 6962-92-1
EINECS: 230-158-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR