4-Iodbutylacetat

Lieferant: CHEMPUR

Synonyme: Essigsäure(4-iodbutyl)ester

RH00804.250 RH00804.1
CHMPRH00804.250EA 0 EUR
CHMPRH00804.250 MAYBRH00804.1
4-Iodbutylacetat
4-Iodbutylacetat
Formel: C₆H₁₁IO₂
Molecular Weight: 242.06 g/mol
MDL: MFCD00010664
CAS-Nummer: 40596-44-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR