4-Iodbutylacetat 96% stabilisiert

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Essigsäure(4-iodbutyl)ester

H31826.09 H31826.18
H31826.09EA 58.6 EUR
H31826.09 H31826.18
4-Iodbutylacetat 96% stabilisiert
4-Iodbutylacetat

Stabilized with copper


Purity: 96% CAS No.: 40596-44-9 Molecular Formula: C6H11IO2 Formula Weight: 242.05 Storage Temperature: 2°C to 8°C Flash Point: 91°(196°F) Refractive Index: 1.5115 Boiling Point: 94-95°/5.5mm MDL No.: MFCD00010664

Formel: C₆H₁₁IO₂
Molecular Weight: 242.06 g/mol
Siedepunkt: 94…95 °C (5.5 mmHg)
Dichte: 1.610
Flammpunkt: 91 °C (196 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00010664
CAS-Nummer: 40596-44-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR