Heptafluorbutyramid

Lieferant: FLUOROCHEM

Synonyme: Perfluorbutyramid, Perfluorbuttersäureamid, Heptafluorbuttersäureamid, 2,2,3,3,4,4,4-Heptafluorbutyramid

006511-5G 006511-25G 006511-10G
FLUO006511-5GEA 39.9 EUR
FLUO006511-5G FLUO006511-25G FLUO006511-10G
Heptafluorbutyramid
Heptafluorbutyramid
Formel: C₄H₂F₇NO
Molecular Weight: 213,05 g/mol
Schmelzpunkt: 105 °C
MDL: MFCD00039773
CAS-Nummer: 662-50-0
EINECS: 211-553-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR