Lanthan(III)sulfat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Scientific

Synonyme: Dilanthantrisulfat Hydrat

43106.22 43106.36
43106.22EA 88.7 EUR
43106.22 43106.36
Lanthan(III)sulfat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lanthan(III)sulfat Hydrat

Purity: 99.9% CAS No.: 57804-25-8 Molecular Formula: La2(SO4)3.xH2O Formula Weight: 566.00(anhy) Physical Form: Crystalline Density: 2.821 Melting Point: dec. white heat Merck Reference: 14,5363 MDL No.: MFCD00011070

Formel: La₂O₁₂S₃·H₂O
Molecular Weight: 584,02 g/mol
Schmelzpunkt: Zers. (Weißglut)
Dichte: 2,821
Lagertemperatur: Raumtemperatur
MDL: MFCD00149760
CAS-Nummer: 57804-25-8
EINECS: 233-239-6
Merck Index: 14,5363

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR